missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Cynomolgus/Rhesus IL-21R Fc Chimera Protein
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
Description
Measured by its binding ability in a functional ELISA. When Recombinant Human IL-21 Protein (Catalog # 8879-IL ) is immoblized at 2.5 μg/mL (100 μL/well), the concentration of Recombinant Cynomolgus Monkey/Rhesus Macaque IL-21R Fc Chimera (Catalog # 10482-R2) that produces a 50% optimal binding response is found to be 0.6-3.6 μg/mL.
Specifications
Specifications
| Accession Number | XP_005591578.1 |
| For Use With (Application) | Bioactivity |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS. |
| Gene ID (Entrez) | 50615 |
| Molecular Weight (g/mol) | M.W.-observed: 60-80 kDa, under reducing conditions, M.W.-theoretical: 52 kDa |
| Quantity | 100 μg |
| Source | Chinese Hamster Ovary cell line, CHO-derived IL-21 R protein Cynomolgus Monkey/Rhesus Macaque IL-21 R (Cys20-Pro236) (N-terminus) Accession # XP_005591578.1 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution. |
| Endotoxin Concentration | <0.10 EU / 1 μg of the protein by the LAL method. |
| Gene Alias | CD360, CD360 antigen, IL-21 R, IL-21 receptor, IL21R, IL-21 R, interleukin 21 receptor, interleukin-21 receptor, MGC10967, NILR, Novel interleukin receptor |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction