missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Cynomolgus Monkey IL-6

Product Code. 18769836
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
This item is not returnable. View return policy

Product Code. 18769836

Brand: R&D Systems™ 10510IL050

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Carrier Free Recombinant Protein

Measured in a cell proliferation assay using T1165.85.2.1 mouse plasmacytoma cells. Nordan, R.P. et al. (1987) J. Immunol. 139:813. The ED50 for this effect is 0.25-1.25 ng/mL.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number NP_001274245.1
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS.
Gene ID (Entrez) 3569
Molecular Weight (g/mol) M.W.-observed: 20-30 kDa, under reducing conditions, M.W.-theoretical: 21 kDa
Quantity 50 μg
Source Human embryonic kidney cell, HEK293-derived cynomolgus monkey IL-6 protein Ala28-Met212
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Gene Alias B-cell differentiation factor, B-cell stimulatory factor 2, BSF2, BSF-2, CDF, CTL differentiation factor, HSF, hybridoma growth factor, IFNB2, IFN-beta-2, IL6, IL-6, Interferon beta-2, interleukin 6 (interferon, beta 2), interleukin BSF-2, interleukin-6, MGI-2 A
Conjugate Unconjugated
Reconstitution Reconstitute at 500 μg/mL in PBS.
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein IL-6
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.