missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Cynomolgus IL-10 R alpha Fc Chimera Protein

Product Code. 18747106
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
This item is not returnable. View return policy

Product Code. 18747106

Brand: R&D Systems™ 10186RI050

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Interleukin-10 Receptor alpha (IL-10 R alpha), also known as IL-10 R1, is a 51 kDa transmembrane glycoprotein member of the class II cytokine receptor family

  • IL-10 R alpha is required for mediating the effects of IL-10, a critical molecule in the control of microbial infections, allergic and autoimmune inflammation, and cancer
  • IL-10 R alpha is the ligand specific subunit of the IL-10 receptor complex
TRUSTED_SUSTAINABILITY

Specifications

Accession Number XP_005579838.1
For Use With (Application) Bioactivity
Gene ID (Entrez) 3587
Molecular Weight (g/mol) 51 kDa
Purification Method >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining
Quantity 50 μg
Source

Human embryonic kidney cell, HEK293-derived cynomolgus monkey IL-10 R alpha protein

Cynomolgus Monkey IL-10 R alpha (His22-Asn235) Accession # XP_005579838.1IEGRMDHuman IgG1 (Pro100-Lys330)N-terminus C-terminus

Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 3 months, -20 to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1 μg of the protein by the LAL method
Conjugate Unconjugated
Reconstitution Reconstitute at 200 μg/mL in PBS
Buffer Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.