missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Cynomolgus Fibroblast Activation Protein alpha
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
Description
Measured by its ability to convert the substrate benzyloxycarbonyl-Gly-Pro-7-amido-4-methylcoumarin (Z-GP-AMC) to Z-Gly-Pro and 7-amino-4-methylcoumarin (AMC). The specific activity is >2500 pmol/min/μg, as measured under the described conditions.
Specifications
Specifications
| Accession Number | XP_005573377 |
| For Use With (Application) | Enzyme Activity |
| Formulation | Supplied as a 0.2 μm filtered solution in Tris, NaCl and Glycerol. |
| Gene ID (Entrez) | 2191 |
| Molecular Weight (g/mol) | M.W. (Observed): 87-94 kDa, under reducing conditions; M.W. (theoretical): 85 kDa |
| Name | 170 kDa melanoma membrane-bound gelatinase, DKFZp686G13158, DPPIV, EC 3.4.21.-, FAP, FAPA, Fibroblast activation protein alpha, fibroblast activation protein, alpha, Integral membrane serine protease, seprase, vibronectin |
| Quantity | 10 μg |
| Source | Chinese Hamster Ovary cell line, CHO-derived cynomolgus monkey Fibroblast Activation Protein alpha/FAP protein Leu26-Asp760 |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 3 months, -20°C to -70°C under sterile conditions after opening. |
| Endotoxin Concentration | <0.10 EU / 1 μg of the protein by the LAL method. |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction