missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Cyno/Rhesus Lysyl Oxidase Homolog 2 Protein, CF
Click to view available options
Quantity:
20 μg
Unit Size:
20µg
Description
Measured by its ability to produce hydrogen peroxide during the oxidation of benzylamine. The specific activity is >6 pmol/min/μg, as measured under the described conditions.
Specifications
Specifications
| Accession Number | XP_005562900.1 |
| Formulation | Supplied as a 0.2 μm filtered solution in MES and NaCl. |
| Gene ID (Entrez) | 4017 |
| Molecular Weight (g/mol) | M.W. (Observed): 92-102 kDa, under reducing conditions, M.W. (Theroretical): 85 kD |
| Quantity | 20 μg |
| Source | Chinese Hamster Ovary cell line, CHO-derived Lysyl Oxidase Homolog 2/LOXL2 protein Gln26-Gln774, with a C-terminal 6-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 3 months, -20°C to -70°C under sterile conditions after opening. |
| Endotoxin Concentration | <0.10 EU per 1 μg of the protein by the LAL method. |
| Gene Alias | EC 1.4.3, EC 1.4.3.-, LOR2, lysyl oxidase homolog 2, lysyl oxidase related 2, lysyl oxidase-like 2, Lysyl oxidase-like protein 2, Lysyl oxidase-related protein 2, Lysyl oxidase-related protein WS9-14, WS9-14 |
| Conjugate | Unconjugated |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction