missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Cyno/Rhesus L-Selectin/CD62L Fc Protein
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
Description
Measured by the ability of the immobilized protein to support the adhesion of LS180 human colorectal adenocarcinoma cells. The ED50 for this effect is 0.300-3.60 μg/mL.
Specifications
Specifications
| Formulation | Supplied as a 0.2 μm filtered solution in PBS with Trehalose. |
| Gene ID (Entrez) | 6402 |
| Molecular Weight (g/mol) | M.W.-observed: 85-95 kDa, under reducing conditions., M.W.-theroretical: 60 kDa |
| Quantity | 100 μg |
| Source | Chinese Hamster Ovary cell line, CHO-derived L-Selectin/CD62L protein Recombinant Cyno/Rhesus L-Selectin (Trp52-Asn345) (C-terminus) Accession # XP_005539996.1 IEGRMD Human IgG1 (Pro100-Lys330) (N-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after opening. 3 months, -20°C to -70°C under sterile conditions after opening. |
| Endotoxin Concentration | <0.10 EU / 1 μg of the protein by the LAL method. |
| Gene Alias | CD62L, CD62L antigen, gp90-MEL, hLHRc, LAM-1, LAM1LECAM1, Leu-8, LEU8, Leukocyte adhesion molecule 1, Leukocyte surface antigen Leu-8, Leukocyte-endothelial cell adhesion molecule 1, LNHRTQ1, LSEL, L-selectin, Lyam-1, LYAM1CD62 antigen-like family member L, Lymph node homing receptor, lymphocyte adhesion molecule 1, pln homing receptor, PLNHR, selectin L |
| Conjugate | Unconjugated |
| Purity or Quality Grade | >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining. |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction