missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Cotton Rat IL-10 (Cys149Tyr) Protein
Description
The Recombinant Cotton Rat IL-10 (Cys149Tyr) Protein is derived from E. coli. The Recombinant Cotton Rat IL-10 (Cys149Tyr) Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
| Formulation | Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. |
| Molecular Weight (g/mol) | 18.7 kDa (monomer) |
| Name | IL-10 (Cys149Tyr) |
| Quantity | 10 μg |
| Source | E. coli-derived cotton rat IL-10 protein Ser1-Ser160 (Cys149Tyr) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 3 months, 2 to 8° C under sterile conditions after reconstitution. |
| Structural Form | Noncovalently-linked homodimer |
| Endotoxin Concentration | <0.1 EU per 1μg of the protein by the LAL method. |
| Gene Alias | CSIF, CSIFMGC126450, Cytokine synthesis inhibitory factor, GVHDS, IL10, IL10A, IL-10MGC126451, interleukin 10, interleukin-10, TGIF |
| Conjugate | Unconjugated |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?