missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Cotton Rat CCL3/MIP-1 alpha Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
Brand: R&D Systems™ 1061-RM-025
Additional Details : Weight : 0.00970kg
Description
The Recombinant Cotton Rat CCL3/MIP-1 alpha Protein is derived from E. coli. The Recombinant Cotton Rat CCL3/MIP-1 alpha Protein has been validated for the following applications: Bioactivity.Specifications
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein. | |
CCL3/MIP-1 alpha | |
E. coli-derived cotton rat CCL3/MIP-1 alpha protein Ala24-Ala92 | |
C-C motif chemokine 3, MIP1-(a), AI323804, chemokine (C-C motif) ligand 3, G0S19-1, LD78a, LD78alpha, MIP1 alpha, MIP-1 alpha, Mip1a, MIP-1alpha, MIP1-alpha, PAT 464.1, SCYA3, SIS-beta | |
Unconjugated | |
97%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
7.9 kDa | |
25μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
CCL3 | |
Recombinant |