missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant CMV CMV Pp65 Protein
A cDNA sequence encoding the CMV Pp65 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11465-500ug
Additional Details : Weight : 0.01000kg
Specifications
Pp65 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
50mM Tris-HCl, 10mM glutathione, 60mM NaCl, 0.125% sarcosil and 50% glycerol. |
500 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
CMV | |
GST | |
CMV Pp65 protein is >95% pure as determined by SDS-PAGE. |