missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Chikungunya Virus CHIKV E2 Protein
A cDNA sequence encoding the CHIKV E2 was constructed and used to recombinantly synthesize the protein.
£173.00 - £1320.00
Specifications
Name | CHIKV E2 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Chikungunya Virus |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15927168
|
enQuireBio™
QP11406-100UG |
100 μg |
£173.00
100µg |
Estimated Shipment: 19-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15947168
|
enQuireBio™
QP11406-500UG |
500 μg |
£660.00
500µg |
Estimated Shipment: 19-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15937168
|
enQuireBio™
QP11406-1MG |
1 mg |
£1320.00
1mg |
Estimated Shipment: 19-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
CHIKV E2 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Chikungunya Virus | |
Untagged | |
Protein is >95% pure as determined by SDS-PAGE. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
Sterile Filtered solution containing PBS and 25mM arginine. |