missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Chicken Netrin-1 Protein

Product Code. 18394747
Change view
Click to view available options
Formulation:
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
Lyophilized from a 0.2μm filtered solution in PBS.
Unit Size:
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Formulation unitSize
18394747 Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. 25µg
18360128 Lyophilized from a 0.2μm filtered solution in PBS. 25µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18394747 Supplier R&D Systems™ Supplier No. 128N1025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Binding Activity

The Recombinant Chicken Netrin-1 Protein is derived from NS0. The Recombinant Chicken Netrin-1 Protein has been validated for the following applications: Binding Activity.
TRUSTED_SUSTAINABILITY

Specifications

Formulation Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
Gene ID (Entrez) 396389
Molecular Weight (g/mol) 66.3 kDa
Name Netrin-1
Quantity 25 μg
Source Mouse myeloma cell line,NS0-derived chicken Netrin-1 protein, (N-terminus) Chicken Netrin-1 (Gly26-Ala606) Accession &Num; Q90922 DIEGRGGGSGGGSGGGS 10-His tag (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.