missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Chicken MDH1 Protein

A cDNA sequence encoding the MDH1 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12665-1000IU

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15941159

  • £99.15 / 1000 units
Estimated Shipment: 14-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

MDH1 Protein
Research Use Only
Recombinant Protein
E. coli
MGEPIRVLVT GAAGQIAYSL LYSIAKGDVF GKEQPLVLVL LDITPMMTVL EGVVMELQDC ALPLLREVIP TDKEEVAFKD LDIAILVGSM PRREGMERKD LLKANVKIFK SQGAALDKYA KKTVKVVVVG NPANTNCLIA SKSAPSIPKE NFSCLTRLDH NRAKSQIALK LGVTSNDVKN VIIWGNHSST QYPDVNHAKV NVKGKEVGVY EAIKDDSWLK GDFILTVQQR GAAVIKARKL SSAMSAAKAI CDHVRDIWFG TPAGEFVSMG VISDGNSYGV PEDLLYSFPV VIKDKTWKFV EGLPINDFSR EKMDLTAKEL TEEKETAVEF LSSA
Greater than 95.0% as determined by(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE.
1000 IU
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Chicken
Untagged
Each mg of protein contains 0.59 mg NaPO4.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers