missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Caronavirus CoV-229E Protein
A cDNA sequence encoding the CoV-229E was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11489-1mg
Additional Details : Weight : 0.01000kg
Specifications
CoV-229E Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
PBS and 3M Urea. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Caronavirus | |
His | |
Protein is >95% pure as determined by 12% PAGE (coomassie staining). |