missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Canine PD-L1/B7-H1 Fc Chimera Protein

Product Code. 18786736
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
This item is not returnable. View return policy

Product Code. 18786736

Brand: R&D Systems™ 10746B7050

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Measured by its binding ability in a functional ELISA. When Recombinant Canine PD-1 Fc Chimera (Catalog # 10553-PD ) is immobilized at 0.5 μg/mL (100 μL/well), Recombinant Canine PD-L1/B7-H1 Fc Chimera (Catalog # 10746-B7) binds with an ED50 of 0.30-4.50 μg/mL.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number NP_001278901.1
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose.
Gene ID (Entrez) 29126
Molecular Weight (g/mol) M.W.-observed: 60-80 kDa, under reducing conditions, M.W.-theroretical: 51 kDa
Quantity 50 μg
Source Human embryonic kidney cell, HEK293-derived canine PD-L1/B7-H1 protein Canine PD-L1/B7-H1 (Phe19-Arg236) (N-terminus) Accession # NP_001278901.1 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Gene Alias Avelumab, B7-H, B7H1, B7-H1, B7H1PDCD1L1, CD274, CD274 antigenMGC142294, CD274 molecule, PDCD1L1, PDCD1LG1, PDCD1LG1MGC142296, PDL1, PD-L1, PD-L1B7 homolog 1, PDL1PDCD1 ligand 1, programmed cell death 1 ligand 1, Programmed death ligand 1
Conjugate Unconjugated
Reconstitution Reconstitute at 500 μg/mL in PBS.
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein PD-L1
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.