missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Canine PD-1 Fc Chimera Protein

Product Code. 30000875
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
This item is not returnable. View return policy

Product Code. 30000875

Brand: R&D Systems™ 10553PD100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Measured by its binding ability in a functional ELISA. When Recombinant Canine PD-1 Fc Chimera (Catalog # 10553-PD) is immobilized at 0.5 μg/mL (100 μL/well), Recombinant Canine PD-L1/B7-H1 Fc Chimera binds with an ED50 of 0.08-6.4 μg/mL.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number NP_001301026.1
Conjugate Unconjugated
For Use With (Application) Bioactivity
Gene ID (Entrez) 5133
Quantity 100 μg
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Synonym CD279, CD279 antigen, hPD-1, PD1, PD1hPD-l, PDCD1, programmed cell death 1, programmed cell death protein 1, Protein PD-1, SLEB2
Target Canine
Buffer Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.