missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Canine IL2 Protein
A cDNA sequence encoding the IL2 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12414-5ug
Additional Details : Weight : 0.01000kg
Specifications
IL2 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSHMAPITS SSTKETEQQM EQLLLDLQLL LNGVNNYENP QLSRMLTFKF YTPKKATEFT HLQCLAEELK NLEEVLGLPQ SKNVHLTDTK ELISNMNVTL LKLKGSETSY NCEYDDETAT ITEFLNKWIT FSQSIFSTLT | |
Greater than 95.0% as determined by SDS-PAGE. |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Canine | |
GST | |
IL2 Canine protein solution (0.5 mg/ml) contains Phosphate buffered saline (pH 7.4). |