missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Canine IL2 Protein

A cDNA sequence encoding the IL2 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12414-1ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15930349

  • £99.15 / 1µg
Estimated Shipment: 12-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

IL2 Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSHMAPITS SSTKETEQQM EQLLLDLQLL LNGVNNYENP QLSRMLTFKF YTPKKATEFT HLQCLAEELK NLEEVLGLPQ SKNVHLTDTK ELISNMNVTL LKLKGSETSY NCEYDDETAT ITEFLNKWIT FSQSIFSTLT
Greater than 95.0% as determined by SDS-PAGE.
1 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Canine
GST
IL2 Canine protein solution (0.5 mg/ml) contains Phosphate buffered saline (pH 7.4).
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers