Learn More
R&D Systems™ Recombinant Canine IFN-gamma Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
Brand:  R&D Systems™ 781-CG-050
Additional Details : Weight : 0.00970kg
 View more versions of this product
Product Code. 18108550
  • £341.24 / 50µg
Estimated Shipment: 17-06-2024
to see stock.
Description
Description
The Recombinant Canine IFN-gamma Protein is derived from E. coli. The Recombinant Canine IFN-gamma Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
Lyophilized from a 0.2μm filtered solution in Sodium Succinate, Mannitol and Tween™ 80 with BSA as a carrier protein.
17 kDa
50μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Recombinant
403801
IFN-gamma
E. coli-derived canine IFN-gamma protein Gln24-Lys166,with an N-terminal Met
Unconjugated
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.