missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Canine HGF Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity, Binding Activity
£345.56
Specifications
Gene ID (Entrez) | 403441 |
---|---|
Name | HGF |
Quantity | 25μg |
Source | Spodoptera frugiperda,Sf 21 (baculovirus)-derived canine HGF protein Arg28-Arg494 (Lys266Gln,Trp321Arg,Leu458Pro) ( alpha chain) & Val495-Ser730 (Ile693Thr) ( beta chain) |
Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
18196350
|
R&D Systems™
3386-HG-025 |
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. |
£345.56
25µg |
Estimated Shipment: 16-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
18333738
|
R&D Systems™
3386-HG-025/CF |
£345.56
25µg |
Estimated Shipment: 16-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The Recombinant Canine HGF Protein is derived from Sf 21 (baculovirus). The Recombinant Canine HGF Protein has been validated for the following applications: Bioactivity, Binding Activity.Specifications
403441 | |
25μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
deafness, autosomal recessive 39, DFNB39, EC 3.4.21, EC 3.4.21.7, fibroblast-derived tumor cytotoxic factor, F-TCF, hepatocyte growth factor (hepapoietin A; scatter factor), Hepatopoeitin-A, Hepatopoietin A, HGFB, HPTA, HPTAhepatocyte growth factor, lung fibroblast-derived mitogen, Scatter factor, SF, SFhepatopoeitin-A | |
Recombinant |
HGF | |
Spodoptera frugiperda,Sf 21 (baculovirus)-derived canine HGF protein Arg28-Arg494 (Lys266Gln,Trp321Arg,Leu458Pro) ( alpha chain) & Val495-Ser730 (Ile693Thr) ( beta chain) | |
Disulfide-linked heterodimer | |
Unconjugated | |
95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |