missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant C. trachomatis Probable outer membrane protein PmpD Protein

A DNA sequence encoding the Chlamydia trachomatis Probable outer membrane protein PmpD, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7502-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16069030

  • £2480.00 / 1mg
Estimated Shipment: 07-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_220332.1
Probable outer membrane protein PmpD Protein
Research Use Only
pmpD
Recombinant Protein
E. coli
EFDYSTNVWG FAFGGFRTLS AENLVAIDGY KGAYGGASAG VDIQLMEDFV LGVSGAAFLG KMDSQKFDAE VSRKGVVGSV YTGFLAGSWF FKGQYSLGET QNDMKTRYGV LGESSASWTS RGVLADALVE YRSLVGPVRP TFYALHFNPY VEVSYASMKF PGFTEQGREA RSFEDASLTN ITIPLGMKFE LAFIKGQFSE VNSLGISYAW EAYRKVEGGA VQLLEAGFDW EGAPMDLPRQ ELRVALENNT EWSSYFSTVL GLTAFCGGFT STDSKLGYEA NTGLRLIF
Greater than 90% as determined by SDS-PAGE.
884611
1 mg
Not determined.
Full Length
C. trachomatis
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers