missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant C. trachomatis Major outer membrane porin Protein
A DNA sequence encoding the Chlamydia trachomatis serovar L2 (strain 434/Bu / ATCC VR-902B) Major outer membrane porin, was expressed in the hosts and tags indicated.
£281.57 - £2480.00
Specifications
Accession Number | WP_009873292.1 |
---|---|
Gene ID (Entrez) | 5858320 |
Name | Major outer membrane porin Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16058030
|
enQuireBio™
QP7292-EC-100UG |
100 μg |
£715.00
100µg |
Estimated Shipment: 27-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16078030
|
enQuireBio™
QP7292-EC-1MG |
1 mg |
£2480.00
1mg |
Estimated Shipment: 27-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16068030
|
enQuireBio™
QP7292-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16088030
|
enQuireBio™
QP7292-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16098030
|
enQuireBio™
QP7292-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16008040
|
enQuireBio™
QP7292-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
WP_009873292.1 | |
Major outer membrane porin Protein | |
Not determined. | |
Full Length | |
C. trachomatis | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
5858320 | |
Research Use Only | |
ompA | |
Recombinant Protein | |
E. coli | |
LPVGNPAEPS LMIDGILWEG FGGDPCDPCT TWCDAISMRM GYYGDFVFDR VLQTDVNKEF QMGAKPTTAT GNAAAPSTCT ARENPAYGRH MQDAEMFTNA AYMALNIWDR FDVFCTLGAT SGYLKGNSAS FNLVGLFGDN ENHATVSDSK LVPNMSLDQS VVELYTDTTF AWSAGARAAL WECGCATLGA SFQYAQSKPK VEELNVLCNA AEFTINKPKG YVGQEFPLDL KAGTDGVTGT KDASIDYHEW QASLALSYRL NMFTPYIGVK WSRASFDADT IRIAQPKSAT TVFDVTTLNP TIAGAGDVKA SAEGQLGDTM QIVSLQLNKM KSRKSCGIAV GTTIVDADKY AVTVETRLID ERAAHVNAQF RF | |
Greater than 90% as determined by SDS-PAGE. |