missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant C. trachomatis 60 kDa chaperonin Protein

Product Code. 16077020
Click to view available options
Quantity:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
200µg
500µg
50µg
This item is not returnable. View return policy

Product Code. 16077020

Brand: enQuireBio™ QP701010ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A DNA sequence encoding the Chlamydia trachomatis (strain D/UW-3/Cx) 60 kDa chaperonin, was expressed in the hosts and tags indicated.

Specifications

Accession Number NP_219613.1
Gene ID (Entrez) 884030
Name 60 kDa chaperonin Protein
Quantity 10 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Gene Symbol groL
Protein Length Full Length
Product Type Recombinant Protein
Cross Reactivity C. trachomatis
Species E. coli
Protein Tag His-SUMO
Sequence MVAKNIKYNE EARKKIQKGV KTLAEAVKVT LGPKGRHVVI DKSFGSPQVT KDGVTVAKEV ELADKHENMG AQMVKEVASK TADKAGDGTT TATVLAEAIY TEGLRNVTAG ANPMDLKRGI DKAVKVVVDQ IRKISKPVQH HKEIAQVATI SANNDAEIGN LIAEAMEKVG KNGSITVEEA KGFETVLDIV EGMNFNRGYL SSYFATNPET QECVLEDALV LIYDKKISGI KDFLPVLQQV AESGRPLLII AEDIEGEALA TLVVNRIRGG FRVCAVKAPG FGDRRKAMLE DIAILTGGQL ISEELGMKLE NANLAMLGKA KKVIVSKEDT TIVEGMGEKE ALEARCESIK KQIEDSSSDY DKEKLQERLA KLSGGVAVIR VGAATEIEMK EKKDRVDDAQ HATIAAVEEG ILPGGGTALI RCIPTLEAFL PMLTNEDEQI GARIVLKALS APLKQIAANA GKEGAIIFQQ VMSRSANEGY DALRDAYTDM LEAGILDPAK VTRSALESAA SVAGLLLTTE ALIAEIPEEK PAAAPAMPGA GMDY
Buffer Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 90% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.