missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant C. botulinum Botulinum neurotoxin type D Protein

A DNA sequence encoding the Clostridium botulinum Botulinum neurotoxin type D, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP9546-ye-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16078451

  • £800.00 / 100µg
Estimated Shipment: 15-01-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

Botulinum neurotoxin type D Protein
Research Use Only
botD
Recombinant Protein
Yeast
MTWPVKDFNY SDPVNDNDIL YLRIPQNKLI TTPVKAFMIT QNIWVIPERF SSDTNPSLSK PPRPTSKYQS YYDPSYLSTD EQKDTFLKGI IKLFKRINER DIGKKLINYL VVGSPFMGDS STPEDTFDFT RHTTNIAVEK FENGSWKVTN IITPSVLIFG PLPNILDYTA SLTLQGQQSN PSFEGFGTLS ILKVAPEFLL TFSDVTSNQS SAVLGKSIFC MDPVIALMHE LTHSLHQLYG INIPSDKRIR PQVSEGFFSQ DGPNVQFEEL YTFGGLDVEI IPQIERSQLR EKALGHYKDI AKRLNNINKT IPSSWISNID KYKKIFSEKY NFDKDNTGNF VVNIDKFNSL YSDLTNVMSE VVYSSQYNVK NRTHYFSRHY LPVFANILDD NIYTIRDGFN LTNKGFNIEN SGQNIERNPA LQKLSSESVV DLFTKVCLRL TK
Greater than 90% as determined by SDS-PAGE.
100 μg
Not determined.
Partial (see sequence information for more details).
C. botulinum
His
Tris-based buffer, 50% glycerol
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers