missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Bovine Thrombin Protein
Click to view available options
Quantity:
100 IU
250 U
50 IU
Unit Size:
100 units
250 units
50 units
Specifications
Specifications
| Name | Thrombin Protein |
| Quantity | 100 IU |
| Regulatory Status | Research Use Only |
| Endotoxin Concentration | < 1.0 EU per ug protein as determined by the LAL method. |
| Biological Activity | 2000 NIH U/mg. |
| Product Type | Recombinant Protein |
| Cross Reactivity | Bovine |
| Species | Bovine |
| Protein Tag | Untagged |
| Buffer | Lyophilized freeze dry powder formulated with Tris buffer and sodium chloride. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction