missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Bovine Placental Lactogen Protein
A cDNA sequence encoding the Placental Lactogen was constructed and used to recombinantly synthesize the protein.
£104.00 - £550.00
Specifications
Name | Placental Lactogen Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | < 1.0 EU per ug protein as determined by the LAL method. |
Biological Activity | Placental Lactogen Bovine is biologically active as evidenced by inducing proliferation of Nb2 cells. |
Product Type | Recombinant Protein |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15925288
|
enQuireBio™
QP10826-1MG |
1 mg |
£550.00
1mg |
Estimated Shipment: 09-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15935288
|
enQuireBio™
QP10826-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15945288
|
enQuireBio™
QP10826-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
Placental Lactogen Protein | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
MAPASSHRGH QWICDLVRGS CLLLLLVVSN LLLCQGVEDY APYCKNQPGN CRIPLQSLFE RATLVASNNY RLAREMFNEF NKQFGEGKNF TSKVINSCHT EFMTTPNNKE AAANTEDEAL LRLVISLLHS WDEPLHQAVT ELLHRNGASP DILARAKEIE DKTKVLLEGV EMIQKRVHPG EKKNEPYPVW SEKSSLTADD EDVRQTAFYR MFHCLHRDSS KISTYINLLK CRFTPC | |
Greater than 98.0% as determined by:(a) Analysis by SEC-HPLC. (b) Analysis by SDS-PAGE. |
Research Use Only | |
Placental Lactogen Bovine is biologically active as evidenced by inducing proliferation of Nb2 cells. | |
Bovine | |
Untagged | |
The protein was lyophilized from a concentrated (1 mg/ml) solution with 0.02-0.03% NaHCO3. |