missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Bovine p85a Protein
A cDNA sequence encoding the p85a was constructed and used to recombinantly synthesize the protein.
£173.00 - £770.00
Spezifikation
Name | p85a Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Bovine |
Produktcode | Marke | Quantity | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Quantity | Preis | Menge & Verfügbarkeit | |||||
15962049
|
enQuireBio™
QP12950-2ug |
2 μg |
£173.00
2µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15942049
|
enQuireBio™
QP12950-10ug |
10 μg |
£439.00
10µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15952049
|
enQuireBio™
QP12950-20ug |
20 μg |
£770.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Spezifikation
p85a Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Bovine | |
Untagged | |
Supplied as a 1 mg/ml solution in 25mM HEPES, pH 8.0, 25mM NaCl, 2.5mM MgCl2 and 50% glycerol. |
Research Use Only | |
Recombinant Protein | |
Leishmania species | |
MSAEGYQYRA LYDYKKEREE DIDLHLGDIL TVNKGSLVAL GFSDGQEAKP EEIGWLNGYN ETTGERGDFP GTYVEYIGRK KISPPTPKPR PPRPLPVAPG PSKTEADSEQ QASTLPDLAE QFAPPDVAPP LLIKLVEAIE KKGLECSTLY RTQSSSNPAE LRQLLDCDTA SLDLEMFDVH VLADAFKRYL LDLPNPVIPV AVSSELISLA PEVQSSEEYI QLLKKLIRSP SIPHQYWLTL QYLLKHFFKL SQTSSKNLLN ARVLSELFSP LLFRFPAASS ENTEHLIKII EILISTEWNE RQPAPALPPK PPKPTTVANN GMNNNMSLQD AEWYWGDISR EEVNEKLRDT ADGTFLVRDA STKMHGDYTL TLRKGGNNKL IKIFHRDGKY GFSDPLTFNS VVELINHYRN ESLAQYNPKL DVKLLYPVSK YQQDQVVKED NIEAVGKKLH EYNTQFQEKS REYDRLYEDY TRTSQEIQMK RTAIEAFNET IKIFEEQCQT QERYSKEYIE KFKREGNETE IQRIMHNYEK LKSRISEIVD SRRRLEEDLK KQAAEYREID KRMNSIKPDL IQLRKTRDQY LMWLTQKGVR QKKLNEWLGN ENTEDQYSLV EDDEDLPHHD EKTWNVGSSN RNKAENLLRG KRDGTFLVRE SSKQGCYACS VVVDGEVKHC VINKTATGYG FAEPYNLYSS LKELVLHYQH TSLVQHNDSL NVTLAYPVYA QQRR | |
Greater than 90.0% as determined by SDS-PAGE. |