missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Bovine IFN-alpha 1 Protein
Shop All Bio Techne Products
Click to view available options
Quantity:
5 μg
Unit Size:
5µg
Specifications
Specifications
| For Use With (Application) | Western Blot, ELISA |
| Formulation | PBS |
| Gene ID (Entrez) | 3439 |
| Quantity | 5 μg |
| Storage Requirements | Store at -20°C. |
| Gene Alias | IFL;IFN;IFNA@;IFNA13;IFN-ALPHA;IFN-alphaD;Interferon alpha-1/13;leIF D |
| Gene Symbol | IFNA1 |
| Reconstitution | Reconstitute with 20 μL sterile PBS containing 0.1% carrier protein. Care should be taken during reconstitution as the protein may appear as a film (at the bottom of the vial. Gently mix vial after reconstitution.) |
| Research Category | Cancer, Immunology, Innate Immunity |
| Purity or Quality Grade | >95%, by SDS-PAGE |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction