missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Bovine GM-CSF Protein

Product Code. 18621918 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18621918 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18621918 Supplier Novus Biologicals™ Supplier No. NBP262475

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

This preparation has been shown to have biological activity in proliferation assays utilizing TF-1 cells.
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Bioactivity
Formulation 1% BSA
Gene ID (Entrez) 1437
Quantity 1 mL
Storage Requirements Store at 4°C. After reconstitution store at -20°C.
Gene Alias colony stimulating factor 2 (granulocyte-macrophage), Colony-stimulating factor, CSF, GM-CSF, GMCSFgranulocyte-macrophage colony-stimulating factor, granulocyte-macrophage colony stimulating factor, MGC131935, MGC138897, molgramostin, sargramostim
Gene Symbol CSF2
Reconstitution Reconstitute with 1.0 mL distilled water. Care should be taken during reconstitution as the protein may appear as a film (at the bottom of the vial. Gently mix vial after reconstitution.)
Inhibitor Properties This preparation has been shown to have biological activity in proliferation assays utilizing TF-1 cells. The specific concentration of GM-CSF has not been assayed.
Research Category Biologically Active Proteins, Cytokine Research, Immunology
Protein GM-CSF
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.