missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Bovine FGF 2 Protein

A cDNA sequence encoding the FGF 2 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP10599-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15954368

  • £320.00 / 10µg
Estimated Shipment: 05-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

281161
10 μg
< 1.0 EU per ug protein as determined by the LAL method.
The recommended concentration in responsive cells is 0.1 to 2ng/ml.
Bovine
Untagged
The FGF-basic Bovine was lyophilized from a concentrated sterile solution containing 50mM Na2HPO4, pH 7.5 and 0.5% HSA.
FGF 2 Protein
Research Use Only
FGF2
Recombinant Protein
Bovine
MAAGSITTLP ALPEDGGSGA FPPGHFKDPK RLYCKNGGFF LRIHPDGRVD GVREKSDPHI KLQLQAEERG VVSIKGVCAN RYLAMKEDGR LLASKCVTDE CFFFERLESN NYNTYRSRKY SSWYVALKRT GQYKLGPKTG PGQKAILFLP MSAKS
Greater than 95.0% as determined by SDS-PAGE.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers