missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Bovine Enteropeptidase/Enterokinase Protein

Product Code. 18379237
Change view
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18379237 10 μg 10µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18379237 Supplier R&D Systems™ Supplier No. 4139SE010

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Measured by its ability to cleave a colorimetric peptide substrate, N-carbobenzyloxy-Lys-ThioBenzyl ester (Z-Lys-SBzl), in the presence of 5,5’Dithio-bis (2-nitrobenzoic acid) (DTNB). Lu, D. et al. (1997) J. Biol. Chem. 272:31293. The specific activity is >35 nmol/min/μg, as measured under the described conditions.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P98072
For Use With (Application) Enzyme Activity
Formulation Supplied as a 0.2 μm filtered solution in Glycerol, NaCl and HEPES.
Gene ID (Entrez) 5651
Molecular Weight (g/mol) M.W. (Observed): 34 kDa, reducing conditions 30 kDa, non-reducing conditions; M.W. (theoretical): 1.5 kDa (heavy chain C-terminal fragment), 26 kDa (light chain)
Name Enteropeptidase/Enterokinase
Quantity 10 μg
Source E. coli-derived bovine Enteropeptidase/Enterokinase protein Cys788-Lys800 (heavy chain C-terminal fragment) with an N-terminal Ala, & Ile801-His1035 (light chain)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 3 months, -20°C to -70°C under sterile conditions after opening.
Structural Form Disulfide-linked heterodimer
Endotoxin Concentration <1.0 EU / 1 μg of the protein by the LAL method.
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade >90%, by SDS-PAGE under reducing conditions and visualized by silver stain
Protein Enteropeptidase/Enterokinase
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.