missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Bovine BTC Protein

A cDNA sequence encoding the BTC was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP10540-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15944188

  • £3965.00 / 1mg
Estimated Shipment: 12-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

280737
1 mg
< 1.0 EU per ug protein as determined by the LAL method.
Recombinant Protein
E. coli
MARAAPGSGA SPLPLLPALA LGLVILHCVV ADGNSTRSPE DDGLLCGDHA ENCPATTTQP KRRGHFSRCP KQYKHYCIKG RCRFVVAEQT PSCVCDEGYA GARCERVDLF YLRGDRGQIL VICLIAVMVI FIILVVSICT CCHPLRKRRK RRKKEEEMET LGKDITPIND DIQETSIA
Greater than 95.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE.
BTC Protein
Research Use Only
BTC
Bovine
Untagged
The Betacellulin Bovine Recombinant was lyophilized after extensive dialysis against 50mM acetic acid.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers