missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Bovine B4GALT1 / GGTB2 Protein
A DNA sequence encoding the Bos taurus (Bovine) B4GALT1 / GGTB2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5702-ec-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_803478.1 | |
B4GALT1 / GGTB2 Protein | |
Research Use Only | |
B4GALT1 | |
Recombinant Protein | |
E. coli | |
MKFREPLLGG SAAMPGASLQ RACR | |
Greater than 90% as determined by SDS-PAGE. |
281781 | |
1 mg | |
Not determined. | |
Cytoplasmic Domain | |
Bovine | |
GST | |
Tris-based buffer, 50% glycerol |