missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Bovine B4GALT1 / GGTB2 Protein

A DNA sequence encoding the Bos taurus (Bovine) B4GALT1 / GGTB2, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP5702-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15947119

  • £715.00 / 100µg
Estimated Shipment: 05-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_803478.1
B4GALT1 / GGTB2 Protein
Research Use Only
B4GALT1
Recombinant Protein
E. coli
MKFREPLLGG SAAMPGASLQ RACR
Greater than 90% as determined by SDS-PAGE.
281781
100 μg
Not determined.
Cytoplasmic Domain
Bovine
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers