missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Bovine Aspartate aminotransferase, mitochondrial Protein

Product Code. 16090850
Click to view available options
Quantity:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
200µg
500µg
50µg
This item is not returnable. View return policy

Product Code. 16090850

Brand: enQuireBio™ QP6108ec500ug

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

A DNA sequence encoding the Bos taurus (Bovine) Aspartate aminotransferase, mitochondrial, was expressed in the hosts and tags indicated.

Specifications

Accession Number NP_777231.1
Gene ID (Entrez) 286886
Name Aspartate aminotransferase, mitochondrial Protein
Quantity 500 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Gene Symbol GOT2
Protein Length Full Length
Product Type Recombinant Protein
Cross Reactivity Bovine
Species E. coli
Protein Tag His-SUMO
Sequence GLAAAASARA SSWWAHVEMG PPDPILGVTE AFKRDTNSKK MNLGVGAYRD DNGKPYVLPS VRKAEAQIAA KNLDKEYLPI AGLAEFCKAS AELALGENNE VLKSGRYVTV QTISGTGALR IGASFLQRFF KFSRDVFLPK PTWGNHTPIF RDAGMQLQSY RYYDPKTCGF DFTGAIEDIS KIPAQSVILL HACAHNPTGV DPRPEQWKEM ATVVKKNNLF AFFDMAYQGF ASGDGNKDAW AVRHFIEQGI NVCLCQSYAK NMGLYGERVG AFTVVCKDAE EAKRVESQLK ILIRPMYSNP PINGARIAST ILTSPDLRKQ WLHEVKGMAD RIISMRTQLV SNLKKEGSSH NWQHIIDQIG MFCYTGLKPE QVERLTKEFS IYMTKDGRIS VAGVTSGNVA YLAHAIHQVT K
Buffer Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 90% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.

Merci de nous aider à améliorer notre site web. Votre commentaire a été soumis