missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant BatCoV RaTG13 Spike S1 Subunit His Protein, CF

Product Code. 18370288
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18370288 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18370288 Supplier R&D Systems™ Supplier No. 10662CV100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Carrier-Free Recombinant Protein

Measured by its binding ability in a functional ELISA with Recombinant Human ACE-2 Fc Chimera (Catalog # 10544-ZN ).
TRUSTED_SUSTAINABILITY

Specifications

Accession Number QHR63300.2
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose.
Molecular Weight (g/mol) M.W. (Observed): 113-127 kDa, under reducing conditions, M.W. (Theoretical): 76 kDa
Quantity 100 μg
Source Chinese Hamster Ovary cell line, CHO-derived batcov ratg13 Spike S1 Subunit protein Val16-Ser680, with a C-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1 μg of the protein by the LAL method.
Gene Alias SARS-CoV-2
Conjugate Unconjugated
Reconstitution Reconstitute at 500 μg/mL in PBS.
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein SARS-CoV-2 Spike S1 Protein
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.