missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Bacteriophage Shiga-like toxin 2 subunit B Protein

A DNA sequence encoding the Enterobacteria phage 933W (Bacteriophage 933W) Shiga-like toxin 2 subunit B, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7305-ye-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16048110

  • £1930.00 / 500µg
Estimated Shipment: 12-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_049501.1
Shiga-like toxin 2 subunit B Protein
Research Use Only
stxB2
Recombinant Protein
Yeast
ADCAKGKIEF SKYNEDDTFT VKVDGKEYWT SRWNLQPLLQ SAQLTGMTVT IKSSTCESGS GFAEVQFNND
Greater than 94.5% as determined by SDS-PAGE.
1262010
500 μg
Not determined.
Full Length
Bacteriophage
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers