missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Bacteria Zinc metalloprotease-1 His Protein

Product Code. 18330136 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18330136 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18330136 Supplier Novus Biologicals™ Supplier No. NBP318169

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

∼0.5U/mg protein Active towards the generic fluorogenic substrate for matrix metalloproteinases MMP2/MMP7
TRUSTED_SUSTAINABILITY

Specifications

Accession Number O53649
For Use With (Application) Enzyme Activity
Formulation 10mM TRIS-HCl, pH 8.0
Molecular Weight (g/mol) M.W. Theoretical: 73.9 kDa
Quantity 50 μg
Storage Requirements Store at -20°C short term. Aliquot and store at -80C long term. Avoid freeze-thaw cycles.
Gene Alias Endopeptidase, peptidase family M13, Zmp1
Research Category Immunology, Innate Immunity
Purity or Quality Grade >98%, by SDS-PAGE
Protein Zinc metalloprotease-1
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.