missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Bacteria NAD Nucleotidase Protein

Product Code. 18377826 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18377826 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18377826 Supplier Novus Biologicals™ Supplier No. NBP318175

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

With AMP: 856 4 nmol of Ade/min/mg of protein With NMN: 110,3 nmol of NR/min/mg of protein With NAD: 5 4 nmol of Ade/min per mg of protein 5 3 nmol of NR/min/mg of protein
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q4QNY4
For Use With (Application) Enzyme Activity
Formulation 5mM KH2PO4, pH 6.8, 100mM TRIS-HCl (pH 8.5) and 10mM MgCl2
Gene ID (Entrez) 56844789
Molecular Weight (g/mol) M.W. Theoretical: 63 kDa
Quantity 50 μg
Storage Requirements Store at -20°C short term. Aliquot and store at -80C long term. Avoid freeze-thaw cycles.
Gene Alias nadN
Gene Symbol nadN
Research Category Immunology, Lipid and Metabolism
Purity or Quality Grade >98%, by SDS-PAGE
Protein NAD Nucleotidase
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.