missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Bacteria Flagellin TLR5 Mutant His Protein

Product Code. 18339256 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18339256 10 μg 10µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18339256 Supplier Novus Biologicals™ Supplier No. NBP318159

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Not detected by human and mouse TLR5 (Toll-like receptor 5). Does not activate TLR5 in human epithelial cell assays based on NF-kB luciferase fusions, in vivo on expression of CCL20 or IL-6. It can be used as negative control for TLR5 signaling assays.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number NP_460912
Concentration LYOPH
For Use With (Application) Bioactivity
Formulation Lyophilized from PBS
Quantity 10 μg
Storage Requirements Store at 4C short term. Aliquot and Store at -20°C long term. Avoid freeze-thaw cycles.
Endotoxin Concentration <0.01 EU/μg purified protein (LAL test; Lonza).
Reconstitution Reconstitute with 100 ul sterile water (0.1mg/ml after reconstitution)
Research Category Immunology, Innate Immunity
Purity or Quality Grade >95% pure by SDS-PAGE
Protein Flagellin
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.