missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Bacillus Monomeric sarcosine oxidase Protein

A DNA sequence encoding the Bacillus sp. (strain NS-129) Monomeric sarcosine oxidase, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7152-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16077490

  • £715.00 / 100µg
Estimated Shipment: 15-01-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

Monomeric sarcosine oxidase Protein
Research Use Only
soxA
Recombinant Protein
E. coli
MSTHFDVIVV GAGSMGMAAG YYLAKQGVKT LLVDSFDPPH TNGSHHGDTR IIRHAYGEGR EYVPFALRAQ ELWYELEKET HHKIFTQTGV LVYGPKGGSA FVSETMEAAN IHSLEHELFE GKQLTDRWAG VEVPDNYEAI FEPNSGVLFS ENCIQAYREL AEAHGATVLT YTPVEDFEVT EDLVTIKTAK GSYTANKLVV SMGAWNSKLL SKLDVEIPLQ PYRQVVGFFE CDEAKYSNNA HYPAFMVEVE NGIYYGFPSF GGSGLKIGYH SYGQQIDPDT INREFGAYPE DEANLRKFLE QYMPGANGEL KKGAVCMYTK TPDEHFVIDL HPKYSNVAIA AGFSGHGFKF SSVVGETLAQ LATTGKTEHD ISIFSLNRDA LKKEAVK
Greater than 90% as determined by SDS-PAGE.
100 μg
Not determined.
Full Length
Bacillus
His
Tris-based buffer, 50% glycerol
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers