missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant B. thetaiotaomicron O-GlcNAcase/OGA Protein

Product Code. 18358917
Change view
Click to view available options
Quantity:
20 μg
Unit Size:
20µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18358917 20 μg 20µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18358917 Supplier R&D Systems™ Supplier No. 6779GH020

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Enzyme Activity

Measured by its ability to hydrolyze 4-methylumbelliferyl-N-acetyl-beta-D-glucosaminide (4-MU-GlcNAc) The specific activity is >3500 pmol/min/μg, as measured under the described conditions.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q89ZI2
For Use With (Application) Enzyme Activity
Formulation Supplied as a 0.2 μm filtered solution in Tris, NaCl, Brij and Glycerol.
Gene ID (Entrez) 10724
Molecular Weight (g/mol) M.W. (Observed): 66-76 kDa, reducing conditions; M.W. (theoretical): 83 kDa
Name O-GlcNAcase/OGA
Quantity 20 μg
Source E. coli-derived b. thetaiotaomicron O-GlcNAcase/OGA protein Gln22-Lys737, with an N-terminal Met and 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 3 months, -20°C to -70°C under sterile conditions after opening.
Endotoxin Concentration <1.0 EU / 1 μg of the protein by the LAL method.
Gene Symbol BT_4395
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade >95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain (at 5 μg / lane)
Protein O-GlcNAcase/OGA/MGEA5
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.