missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant B. thetaiotaomicron Heparinase I Protein

Product Code. 18308997
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
This item is not returnable. View return policy

Product Code. 18308997

Brand: R&D Systems™ 5830GH010

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Enzyme Activity

Measured by its ability to liberate oligosaccharides from heparin. The specific activity is >12,000 pmol/min/μg, as measured under the described conditions.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number NP_813586
For Use With (Application) Enzyme Activity
Formulation Supplied as a 0.2 μm filtered solution in Tris, NaCl and Citrate.
Gene ID (Entrez) 1073402
Molecular Weight (g/mol) M.W. (Observed): 59 kDa, reducing conditions; M.W. (theoretical): 55 kDa
Name Heparinase I
Quantity 10 μg
Source E. coli-derived b. thetaiotaomicron Heparinase I protein Met1-Arg376, with Met, 6-His tag and a fusion partner at the N-terminus
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -70°C as supplied. 3 months, -70°C under sterile conditions after opening.
Endotoxin Concentration <1.0 EU / 1 μg of the protein by the LAL method.
Gene Symbol BT_4675
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade >90%, by SDS-PAGE under reducing conditions and visualized by silver stain
Protein Heparinase I
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.