missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant B. subtilis NAPRT1 Protein

Product Code. 18393504 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18393504 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18393504 Supplier Novus Biologicals™ Supplier No. NBP318170

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

∼0.8 U/mg protein One unit is defined as the amount of enzyme that synthesizes 1umol nicotinic acid mononucleotide (NaMN) per min
TRUSTED_SUSTAINABILITY

Specifications

Accession Number O32090
For Use With (Application) Enzyme Activity
Formulation 20mM TRIS-HCl, pH 7.8, 50mM NaCl
Gene ID (Entrez) 93100
Molecular Weight (g/mol) M.W. Theoretical: 56 kDa
Quantity 50 μg
Storage Requirements Store at -20°C short term. Aliquot and store at -80C long term. Avoid freeze-thaw cycles.
Gene Alias EC 2.4.2.11, FHA-HIT-interacting protein, FHIP, NAPRTase, nicotinate phosphoribosyltransferase, nicotinate phosphoribosyltransferase domain containing 1, Nicotinate phosphoribosyltransferase domain-containing protein 1, nicotinic acid phosphoribosyltransferase, PP3856
Gene Symbol NAPRT
Research Category Lipid and Metabolism
Purity or Quality Grade >98%, by SDS-PAGE
Protein NAPRT1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.