missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant B. subtilis NADK Protein

Product Code. 18359994 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18359994 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18359994 Supplier Novus Biologicals™ Supplier No. NBP318171

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

∼2 6 U/mg protein One unit is defined as the amount of enzyme that synthesizes 1umol nicotinamide adenine dinucleotide phosphate (NADP) per min
TRUSTED_SUSTAINABILITY

Specifications

Accession Number O31612
For Use With (Application) Enzyme Activity
Formulation 50mM TRIS-HCl, pH 7.0, 150mM NaCl, 1mM EDTA and 1mM dithiothreitol
Gene ID (Entrez) 65220
Molecular Weight (g/mol) M.W. Theoretical: 30 kDa
Quantity 50 μg
Storage Requirements Store at -20°C short term. Aliquot and store at -80C long term. Avoid freeze-thaw cycles.
Gene Alias dJ283E3.1, DKFZp686L22239, EC 2.7.1, EC 2.7.1.23, FLJ13052, FLJ37724, FLJ54695, FLJ77769, FLJ78247, FLJ78307, MGC1900, NAD kinase, Poly(P)/ATP NAD kinase
Gene Symbol NADK
Research Category Cancer, Immunology, Lipid and Metabolism
Purity or Quality Grade >98%, by SDS-PAGE
Protein NADK
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.