missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Bacterial leucyl aminopeptisdase protein sequentially cleaves N-terminal amino acids except E, D, and X-P.
Specifications
Specifications
| Concentration | LYOPH |
| For Use With (Application) | PAGE, Bioactivity |
| Formulation | Lyophilized from a 0.2 um filtered concentrated solution in 20 mM Tris, 150 mM NaCl, 5uM ZnSO4, pH 8.0. |
| Molecular Weight (g/mol) | 31.4 kDa |
| Quantity | 1 mg |
| Storage Requirements | Store at -20°C to -70°C as supplied. After reconstitution, store at 2°C to 8°C for 1 month and at -20°C to -70°C for long term storage. Avoid repeated freeze-thaw cycles. |
| Endotoxin Concentration | Less than 0.1 EU/ug of Aeromonas Aminopeptidase as determined by LAL method. |
| Gene Alias | Aeromonas Aminopeptidase, Aminopeptidase, Bacterial leucyl aminopeptidase, EC 3.4.11.10., Vibrio proteolyticus (Aeromonas proteolytica) |
| Reconstitution | Recommended to centrifuge prior to opening. Reconstitute in sterile distilled water or aqueous buffer to a concentration of 0.1-1.0mg/mL. Apportion stock solutions into working aliquots and store at <-20°C. |
| Research Category | Virology Bacteria and Parasites |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?