missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant A. victoria Aequorea victoria GFP Protein

A DNA sequence encoding the Aequorea victoria (Jellyfish) Aequorea victoria GFP, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7182-ye-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16007600

  • £800.00 / 100µg
Estimated Shipment: 27-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

GFP Protein
Research Use Only
GFP
Recombinant Protein
Yeast
MSKGEELFTG VVPILVELDG DVNGHKFSVS GEGEGDATYG KLTLKFICTT GKLPVPWPTL VTTFSYGVQC FSRYPDHMKQ HDFFKSAMPE GYVQERTIFF KDDGNYKTRA EVKFEGDTLV NRIELKGIDF KEDGNILGHK LEYNYNSHNV YIMADKQKNG IKVNFKIRHN IEDGSVQLAD HYQQNTPIGD GPVLLPDNHY LSTQSALSKD PNEKRDHMVL LEFVTAAGIT HGMDELYK
Greater than 90% as determined by SDS-PAGE.
100 μg
Not determined.
Full Length
A. victoria
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers