missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant A. Muciniphila M60 domain-containing peptidase
Click to view available options
:
20 μg
Beschreibung
Measured by its ability to digest MUC16. >90% of MUC16 is digested by rAm M60 peptidase, as measured under the described conditions.
Spezifikation
Spezifikation
| Accession Number | WP_012419679.1 |
| For Use With (Application) | Enzyme Activity |
| Formulation | Supplied as a 0.2 μm filtered solution in Tris and NaCl. |
| Molecular Weight (g/mol) | MolecularWeight-observed: 52-57 kDa, under reducing conditions, MolecularWeight-theroretical: 56 kDa |
| Quantity | 20 μg |
| Source | E. coli-derived akkermansia muciniphila M60 domain-containing protein Ala21-Glu506 with an N-terminal Met and C-terminal 6-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70°C as supplied. 3 months, -20 to -70°C under sterile conditions after opening. |
| Endotoxin Concentration | <1.0 EU per 1 μg of the protein by the LAL method. |
| Gene Alias | Akkermansia muciniphila ATCC BAA-835 Peptidase M60, Akkermansia muciniphila ATCC BAA-835 Peptidase M60 domain-containing protein, Amuc_0627, conserved hypothetical protein [Akkermansia mucini, conserved hypothetical protein [Akkermansia muciniphila ATCC BAA-835], M60 family metallopeptidase [Akkermansia muciniphi, M60 family metallopeptidase [Akkermansia muciniphila] |
| Conjugate | Unconjugated |
| Mehr anzeigen |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur