missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RECK Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£390.00
Specifications
| Antigen | RECK |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
RECK Polyclonal specifically detects RECK in Mouse samples. It is validated for Western Blot.Specifications
| RECK | |
| Polyclonal | |
| Rabbit | |
| NP_057887 | |
| 8434 | |
| The immunogen for this antibody is Reck. Peptide sequence SCIVGGKRKSHGTSFTIDCNVCSCFAGNLVCSTRLCLSEHSSDDDRRTFT. | |
| Primary |
| Western Blot | |
| Unconjugated | |
| RUO | |
| hRECK, membrane-anchored glycoprotein (metastasis and invasion), reversion-inducing-cysteine-rich protein with kazal motifs, ST15reversion-inducing cysteine-rich protein with Kazal motifs, suppression of tumorigenicity 15 (reversion-inducing-cysteine-rich protein withkazal motifs), suppression of tumorigenicity 5 (reversion-inducing-cysteine-rich protein withkazal motifs), Suppressor of tumorigenicity 15 protein | |
| RECK | |
| IgG | |
| 104 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title