missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
RECK Monoclonal antibody specifically detects RECK in Human samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | RECK |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | MM0523-9G19 |
| Conjugate | Janelia Fluor 646 |
| Dilution | Western Blot |
| Formulation | 50mM Sodium Borate |
| Gene Alias | hRECK, membrane-anchored glycoprotein (metastasis and invasion), reversion-inducing-cysteine-rich protein with kazal motifs, ST15reversion-inducing cysteine-rich protein with Kazal motifs, suppression of tumorigenicity 15 (reversion-inducing-cysteine-rich protein withkazal motifs), suppression of tumorigenicity 5 (reversion-inducing-cysteine-rich protein withkazal motifs), Suppressor of tumorigenicity 15 protein |
| Host Species | Mouse |
| Immunogen | Human recombinant RECK |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?