missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RECK Antibody (MM0523-9G19), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-11863F
This item is not returnable.
View return policy
Description
RECK Monoclonal antibody specifically detects RECK in Human samples. It is validated for Western Blot
Specifications
| RECK | |
| Monoclonal | |
| FITC | |
| PBS | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| MM0523-9G19 | |
| Western Blot | |
| hRECK, membrane-anchored glycoprotein (metastasis and invasion), reversion-inducing-cysteine-rich protein with kazal motifs, ST15reversion-inducing cysteine-rich protein with Kazal motifs, suppression of tumorigenicity 15 (reversion-inducing-cysteine-rich protein withkazal motifs), suppression of tumorigenicity 5 (reversion-inducing-cysteine-rich protein withkazal motifs), Suppressor of tumorigenicity 15 protein | |
| Human recombinant RECK | |
| 0.1 mL | |
| Cell Cycle and Replication | |
| 8434 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction